4-tert-Butyl-N-(3-fluorobenzyl)aniline, 97%
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
H57772.MDEA 276 DKK
ALFAH57772.MD ALFAH57772.03
4-tert-Butyl-N-(3-fluorobenzyl)aniline, 97%
Purity: 97%
CAS No.: 1036463-19-0
Molecular Formula: C17H20FN
Formula Weight: 257.35
MDL No.: MFCD12616324