3-Chloro-N-(4-fluorobenzyl)-2-methylaniline, 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

H57117.03EA 3300 DKK
ALFAH57117.03 ALFAH57117.MD
3-Chloro-N-(4-fluorobenzyl)-2-methylaniline, 97%
Purity: 97%
CAS No.: 356531-60-7
Molecular Formula: C14H13ClFN
Formula Weight: 249.71
MDL No.: MFCD03210775
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR