4-Bromo-N-(2-fluorobenzyl)-2-methylaniline, 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

H57474.MDEA 247 DKK
ALFAH57474.MD ALFAH57474.03
4-Bromo-N-(2-fluorobenzyl)-2-methylaniline, 97%
Purity: 97%
CAS No.: 1019603-65-6
Molecular Formula: C14H13BrFN
Formula Weight: 294.16
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR