3-Iodophenylsulphur pentafluoride ≥93.0% (by GC)
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
TCIAI0694-1GEA
1020
DKK
TCIAI0694-1G
3-Iodophenylsulphur pentafluoride ≥93.0% (by GC)
3-Iodophenylsulphur pentafluoride
Formel:
C₆H₄F₅IS Kogepunkt: 57°C Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD03788513 CAS-nummer: 286947-67-9 |
Specifikationer Testresultater
Appearance | Colorless to Red to Green clear liquid |
Purity (GC) | min. 93.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...