5-Amino-2,4,6-triiodoisophthalic acid ≥96.0% (by HPLC, titration analysis)

Leverandør: TCI EUROPE
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

TCIAA1406-25GEA 542 DKK
TCIAA1406-25G
5-Amino-2,4,6-triiodoisophthalic acid ≥96.0% (by HPLC, titration analysis)
5-Amino-2,4,6-triiodoisophthalic acid
Formel: C₈H₄I₃NO₄
Molekylvægt: 558,84
MDL Number: MFCD00190167
CAS-nummer: 35453-19-1
EINECS: 252-575-4
https://dk.vwr.com/stibo/web/std.lang.all/93/58/28929358.jpg

Bestil nu

Specifikationer Testresultater

Appearance White to Gray to Brown powder to crystal
Purity (HPLC) min. 96.0 area%
Purity (Neutralization titration) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR