4-Fluorophenylsulphur pentafluoride ≥95.0% (by GC)
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
TCIAF0824-1GEA
1140
DKK
TCIAF0824-1G
4-Fluorophenylsulphur pentafluoride ≥95.0% (by GC)
4-Fluorophenylsulphur pentafluoride
Formel:
C₆H₄F₆S Molekylvægt: 222,15 g/mol Kogepunkt: 103°C Flammepunkt: 48°C Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD16817601 CAS-nummer: 1063625-86-4 UN nummer: 1993 ADR: 3,III |
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...