Potassium 3-fluoro-4-methoxyphenyltrifluoroborate, 96%
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
H53060.03EA 812 DKK
ALFAH53060.03 ALFAH53060.MD
Potassium 3-fluoro-4-methoxyphenyltrifluoroborate, 96%
Purity: 96%
CAS No.: 850623-62-0
Molecular Formula: C7H6BF4KO
Formula Weight: 232.03
Melting Point: 276-280°
MDL No.: MFCD04115757