Ethyl phenoxyacetate ≥98.0% (by GC)
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
TCIAP1340-25GEA
977
DKK
TCIAP1340-25G
Ethyl phenoxyacetate ≥98.0% (by GC)
Ethyl phenoxyacetate
Formel:
C₁₀H₁₂O₃ Molekylvægt: 180,20 g/mol |
MDL Number:
MFCD00026895 CAS-nummer: 2555-49-9 EINECS: 219-867-3 |
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...