Phytol (mixture of cis and trans isomers) ≥90.0% (by GC)
Leverandør: TCI EUROPE
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
TCIAP0411-25GEA 755 DKK
TCIAP0411-25G
Phytol (mixture of cis and trans isomers) ≥90.0% (by GC)
Phytol (mixture of cis and trans isomers)
Formel: C₂₀H₄₀O
MDL Number: MFCD00151280
CAS-nummer: 150-86-7
EINECS: 205-776-6
https://dk.vwr.com/stibo/web/std.lang.all/11/08/28991108.jpg
Bestil nu
Specifikationer Testresultater
Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 90.0 %