1-Bromo-3-(trimethylsilyl)benzene ≥96.0% (by GC)
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
TCIAB3477-5GEA
788
DKK
TCIAB3477-5G
1-Bromo-3-(trimethylsilyl)benzene ≥96.0% (by GC)
1-Bromo-3-(trimethylsilyl)benzene
Formel:
C₉H₁₃BrSi Molekylvægt: 229,1890 g/mol Kogepunkt: 50°C |
MDL Number:
MFCD00093882 CAS-nummer: 17878-47-6 |
Specifikationer Testresultater
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...