Methyldiphenylphosphine oxide ≥98.0% (by GC)
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
TCIAD2445-10GEA
600
DKK
TCIAD2445-10G
Methyldiphenylphosphine oxide ≥98.0% (by GC)
Methyldiphenylphosphine oxide
Formel:
C₁₃H₁₃OP Molekylvægt: 216,22 g/mol Kogepunkt: 180°C |
MDL Number:
MFCD00002081 CAS-nummer: 2129-89-7 EINECS: 218-346-8 |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 111.0 to 115.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...