3,5-Dimethyl-1H-pyrazole-1-methanol ≥98.0% (by titrimetric analysis)

Leverandør: TCI EUROPE
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

TCIAD2199-25GEA 501 DKK
TCIAD2199-25G
3,5-Dimethyl-1H-pyrazole-1-methanol ≥98.0% (by titrimetric analysis)
3,5-Dimethyl-1H-pyrazole-1-methanol
Formel: C₆H₁₀N₂O
MDL Number: MFCD00020738
CAS-nummer: 85264-33-1
EINECS: 286-553-0
https://dk.vwr.com/stibo/web/std.lang.all/90/55/28979055.jpg

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 108.0 to 112.0 deg-C
Solubility in hot Toluene almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR