Dicyclohexylchlorophosphine ≥97.0% (by titrimetric analysis)

Leverandør: TCI EUROPE
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

TCIAC1658-5GEA 1830 DKK
TCIAC1658-5G TCIAC1658-1G
Dicyclohexylchlorophosphine ≥97.0% (by titrimetric analysis)
Dicyclohexylchlorophosphine
Formel: C₁₂H₂₂ClP
Molekylvægt: 232,73 g/mol
Kogepunkt: 166°C
MDL Number: MFCD00014294
CAS-nummer: 16523-54-9
UN nummer: 3265
ADR: 8,II
https://dk.vwr.com/stibo/web/std.lang.all/88/72/28938872.jpg

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 90.0 %
Purity (Argentmetric Titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR