p-Anisidin 0,25% in anhydrous acetic acid

Leverandør: Ampliqon A/S
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering


Synonymer: 4-Methoxyanilin4-Anisidin

AMPQ10430.0100EA 3970 DKK
AMPQ10430.0100
p-Anisidin 0,25% in anhydrous acetic acid
p-Anisidin
Formel: C₇H₉NO
Molekylvægt: 123,15 g/mol
MDL Number: MFCD00007864
CAS-nummer: 104-94-9
EINECS: 203-254-2
UN nummer: 2789
ADR: 8,II

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR