1,5-Diamino-4,8-dinitro-9,10-anthraquinone ≥96.0% (by titrimetric analysis)
Leverandør: TCI EUROPE
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
TCIAD2727-25GEA 2080 DKK
TCIAD2727-25G TCIAD2727-5G
1,5-Diamino-4,8-dinitro-9,10-anthraquinone ≥96.0% (by titrimetric analysis)
1,5-Diamino-4,8-dinitro-9,10-anthraquinone
Formel: C₁₄H₆N₂O₈
MDL Number: MFCD00035827
CAS-nummer: 128-91-6
EINECS: 204-918-4
UN nummer: 0000
https://dk.vwr.com/stibo/web/std.lang.all/98/46/28979846.jpg
Bestil nu
Specifikationer Testresultater
Appearance White to Yellow to Orange powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (Neutralization titration) min. 96.0 %