N-(4-Fluorobenzyl)-3,5-dimethylaniline, 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

H57120.03EA 1660 DKK
ALFAH57120.03 ALFAH57120.MD
N-(4-Fluorobenzyl)-3,5-dimethylaniline, 97%
Purity: 97%
CAS No.: 356531-58-3
Molecular Formula: C15H16FN
Formula Weight: 229.30
MDL No.: MFCD03210773
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR