3-Methylbenzothiazolium iodide ≥98.0% (by titrimetric analysis)

Leverandør: TCI EUROPE
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

TCIAM1131-25GEA 1320 DKK
TCIAM1131-25G
3-Methylbenzothiazolium iodide ≥98.0% (by titrimetric analysis)
3-Methylbenzothiazolium iodide
Formel: C₈H₈INS
MDL Number: MFCD00050297
CAS-nummer: 2786-31-4
EINECS: 220-505-1
https://dk.vwr.com/stibo/web/std.lang.all/48/28/28974828.jpg

Bestil nu

Specifikationer Testresultater

Appearance White to Light red to Green powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in hot Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR