3-(Carboxymethyl)benzothiazolium bromide ≥97.0% (by titrimetric analysis)

Leverandør: TCI EUROPE
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

TCIAC1496-25GEA 583 DKK
TCIAC1496-25G
3-(Carboxymethyl)benzothiazolium bromide ≥97.0% (by titrimetric analysis)
3-(Carboxymethyl)benzothiazolium bromide
Formel: C₉H₈BrNO₂S
MDL Number: MFCD00134360
CAS-nummer: 74385-09-4
UN nummer: 0000
https://dk.vwr.com/stibo/web/std.lang.all/87/15/28938715.jpg

Bestil nu

Specifikationer Testresultater

Appearance White to Orange to Green powder to crystal
Purity (Neutralization titration) min. 97.0 %
Solubility in hot Water very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR