3-Ethylbenzothiazolium bromide ≥98.0% (by HPLC, titration analysis)
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
TCIAE0395-10GEA
788
DKK
TCIAE0395-10G
3-Ethylbenzothiazolium bromide ≥98.0% (by HPLC, titration analysis)
3-Ethylbenzothiazolium bromide
Formel:
C₉H₁₀BrNS Molekylvægt: 244,16 g/mol |
MDL Number:
MFCD00051852 CAS-nummer: 32446-47-2 |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...