5-Chloro-3-phenyl-2,1-benzisoxazole ≥96.0% (by GC)

Leverandør: TCI EUROPE
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

TCIAC1687-25GEA 156 DKK
TCIAC1687-25G
5-Chloro-3-phenyl-2,1-benzisoxazole ≥96.0% (by GC)
5-Chloro-3-phenyl-2,1-benzisoxazole
Formel: C₁₃H₈ClNO
Molekylvægt: 229,67
MDL Number: MFCD00014573
CAS-nummer: 719-64-2
EINECS: 211-950-2
https://dk.vwr.com/stibo/web/std.lang.all/88/96/28938896.jpg

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 96.0 %
Melting point 114.0 to 117.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR