4-Methylstilbene ≥98.0% (by GC)

Leverandør: TCI EUROPE
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

TCIAM1492-5GEA 2630 DKK
TCIAM1492-5G
4-Methylstilbene ≥98.0% (by GC)
4-Methylstilbene
Formel: C₁₅H₁₄
MDL Number: MFCD00452726
CAS-nummer: 4714-21-0
https://dk.vwr.com/stibo/web/std.lang.all/57/47/28975747.jpg

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 118.0 to 122.0 deg-C
Solubility in Toluene almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR