Reinecke's salt monohydrate
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
TCIAR0004-25GEA
353
DKK
TCIAR0004-25G
Reinecke's salt monohydrate
Reinecke's salt monohydrate
Formel:
C₄H₁₀CrN₇S₄·1H₂O Molekylvægt: 354,41 g/mol |
MDL Number:
MFCD00150925 CAS-nummer: 13573-17-6 EINECS: 237-003-3 |
Specifikationer Testresultater
Appearance | Orange to Brown to Dark purple powder to crystal |
Absorbance | min. 0.400 (2 g/L, H2O, 518.0 to 522.0 nm) |
Sensitiveness | to pass test |
Precipitate capacity | to pass test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...