Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')praseodymium(III) ≥98.0% (by titrimetric analysis) NMR shift reagent

Leverandør: TCI EUROPE
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

TCIAT1264-1GEA 1040 DKK
TCIAT1264-1G
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')praseodymium(III) ≥98.0% (by titrimetric analysis) NMR shift reagent
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')praseodymium(III)
Formel: C₃₃H₅₇O₆Pr
Molekylvægt: 690,71 g/mol
MDL Number: MFCD00000027
CAS-nummer: 15492-48-5
EINECS: 239-522-0
https://dk.vwr.com/stibo/web/std.lang.all/53/04/28995304.jpg

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow to Green powder to crystal
Purity (Chelometric Titration) min. 98.0 %
Melting point 219.0 to 221.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR