Pipette controller, manual, Profiller™ 435
Leverandør: Socorex
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Profiller™
SOCO435.100EA 620 DKK
SOCO435.100
Pipette controller, manual, Profiller™ 435
Pipetter Pipettehjælpere
This manual, easy to use pipette controller provides smooth control in both aspirating and dispensing, using glass or plastic pipettes of volume up to 100 ml.
  • Large capacity bulb
  • Soft action lever button and efficient blow out system
  • PTFE protection membrane filter
  • Maintenance free
  • Alternative colour coded nose cones available