Measuring cup
Leverandør: Vitlab
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
VITL69394EA 150 DKK
VITL69394
Measuring cup
PP, translucent.
  • Good temperature resistance
  • Good chemical resistance
Ideal for general laboratory use and a wide variety of other applications. With embossed scale.