Isolator gloves, neoprene
Leverandør: Piercan
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
PIECE10330CN7510U1EA 552 DKK
PIECE10330CN7510U1 PIECE10330CNS8E5U1 PIECE10330CNS7E3U1
Isolator gloves, neoprene
Handsker
These gloves, made of non staining neoprene, provide good mechanical protection and good resistance against various chemicals.
These gloves, made of non staining neoprene, provide good mechanical protection and good resistance against various chemicals. Ideal for use in life science applications (pharmaceutical and medical).
  • Good behavior against UV and ozone
  • Excellent dexterity and flexibility
  • Excellent resistance to sterilisation processes (gamma and beta radiation)
Ideal for use in life science applications (pharmaceutical and medical).
  • Good behavior against UV and ozone
  • Excellent dexterity and flexibility
  • Excellent resistance to sterilisation processes (gamma and beta radiation)
0,3 mm thickness: EN 374-1, EN 421; Cat. III
0,5 mm thickness: EN 388 (X100), EN 374 (ACK), EN 421; Cat. III
Certificeringer: EN ISO 374-1 Type B (ANP), EN ISO 374-5 VIRUS, EN 421; Cat. III