Graduated pipettes, BLAUBRAND®, USP, type 2, Class AS, total delivery
Leverandør: Brand
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BLAUBRAND® ETERNA
BRNDUSP27513EA 359 DKK
BRNDUSP27513 BRNDUSP27511 BRNDUSP27506 612-4755 612-4754 612-4757 BRNDUSP27509 612-4756
Graduated pipettes, BLAUBRAND®, USP, type 2, Class AS, total delivery
Pipetter Pipetter med inddeling
AR-Glas®
  • Calibrated to deliver (TD, Ex), nominal volume at the top
  • Marks and inscriptions in high contrast blue enamel
  • Waiting time 5 seconds
  • Suitable for cotton plugs
  • Incl. USP batch certificate, on request also available with USP individual certificate
DIN EN ISO 835
Length: 360 ±10 mm
Certificeringer: DIN EN ISO 835. DE-M marking.
Advarsel: * Error limit USP: ±0,01 ml.
* Error limit USP: ± 0,01 ml