Graduated pipettes, SILBERBRAND ETERNA, type 2, Class B, total delivery
Leverandør: Brand
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
SILBERBRAND ETERNA
BRND27009EA 630 DKK
BRND27009 BRND27005 BRND27006 BRND27013 BRND27002 BRND27010 BRND27011
Graduated pipettes, SILBERBRAND ETERNA, type 2, Class B, total delivery
Pipetter Pipetter med inddeling
AR-Glas®.
  • Calibrated to deliver (TD, Ex)
  • Nominal volume at the top
DIN EN ISO 835
Length: 360 mm ±10 mm except for 25 ml pipettes: 450 mm ±10 mm.
Certificeringer: DIN EN ISO 835
Advarsel: * Suitable for cotton plugs
* Suitable for cotton plugs