Capillary blood collection tubes, MiniCollect®, Vacuette®
Leverandør: Greiner bio-one
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
MiniCollect®
GREI450412_1000EA 930 DKK
GREI450412_1000 GREI450472BE GREI450474DE 720-2784 GREI450472_100 GREI450479BE_U GREI450475BE GREI450474_1000 GREI450477_1000 GREI450475_100 GREI450477_100 GREI450481_1000 GREI450409_1000 GREI450470_1000 GREI450472_1000 GREI450475_1000 GREI450471_100
Capillary blood collection tubes, MiniCollect®, Vacuette®
Kliniske diagnose systemer Clinical Sample Collection/Preparation
PP, translucent, with cap.
  • With or without additives
MiniCollect® tubes are ideal for collecting, transporting and processing capillary blood. Capillary blood collection is especially recommended in patients with fragile veins, young children, geriatric patients and in situations where only a small blood sample is required. MiniCollect® tubes are available with a volume of 0,25 to 1 ml and with a range of additives.
Advarsel: *Produktet/produkterne på denne side er uden for anvendelsesområdet for EU -direktiv 2019/904