Combined reference electrode, XM950
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
HACHB31M950EA 2240 DKK
HACHB31M950
Combined reference electrode, XM950
Silver electrode for the determination of mercaptan by titration.