Capillary columns, VF-1701ms
Leverandør: Agilent Technologies
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
VARICP9148EA 6860 DKK
VARICP9148 568-0769 568-0758 568-0759 568-0756 568-0767 568-0757 568-0768 568-0765 568-0766 568-0763 568-0764 568-0761 568-0762 568-0770 568-0760
Capillary columns, VF-1701ms
Kromatografikolonner
GC column for pesticides, PCBs and semi-volatile organic compounds.
  • Highly inert for difficult analytes such as p,p'-DDT to improve productivity
  • Column deactivation for more accurate trace analysis
  • Eliminate ghost peaks and unstable baselines for more reliable data
Ultra-low bleed 14% cyanopropyl/phenyl, 86% PDMS.
Temperature limits: −20 to +280/300 °C.
Similar phases: SPB-1701, Rtx-1701, BP-10, OV-1701, 007-1701, ZB-1701.